missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CCL2 (MCP-1) Recombinant Protein, PeproTech®

Artikelnummer. 17871483
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
2 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17871483 250 μg 250 Mikrogramm
17841493 100 μg 100 Mikrogramm
17841193 10 μg 10 Mikrogramm
17800833 1 mg 1 Milligramm
17890813 2 μg 2 Mikrogramm
17830643 500 μg 500 Mikrogramm
17840523 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17871483 Lieferant Gibco™ Lieferanten-Nr. 25010250UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Recombinant Murine JE is a 13.8 kDa protein containing 125 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P10148
Zur Verwendung mit (Anwendung) ELISA, Functional Assay, Western Blot
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 20296
Molekulargewicht 13.8 kDa
Name Mouse CCL2 (MCP-1)
Menge 250 μg
Quelle E. coli
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1 A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1 A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Gebräuchliche Bezeichnung CCL2 (MCP-1)
Gensymbol Ccl2
Biologische Aktivität Determined by its ability to chemoattract Balb/C mouse spleen MNCs using a concentration range of 1.0-20.0 ng/ml
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz QPDAVNAPLT CCYSFTSKMI PMSRLESYKR ITSSRCPKEA VVFVTKLKRE VCADPKKEWV QTYIKNLDRN QMRSEPTTLF KTASALRSSA PLNVKLTRKS EANASTTFST TTSSTSVGVT SVTVN
Inhalt und Lagerung -20°C
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.