Learn More
R&D Systems™ Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit, R&D Systems™
Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich 4.70 - 300pg/mL
Assay-Empfindlichkeit: 1.5pg/mL
Probentyp Cell Culture Supernatant, Serum
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL3/MIP-1 alpha in cell culture supernates (50 ul), serum (50 ul).
Spezifikation
Spezifikation
4.70 - 300pg/mL
1.5pg/mL
Cell Culture Supernatant, Serum