Learn More
R&D Systems™ Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit, R&D Systems™
Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 4.70 - 300pg/mL |
---|---|
Assay-Empfindlichkeit: | 1.5pg/mL |
Probentyp | Cell Culture Supernatant, Serum |
Beschreibung
The Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL3/MIP-1 alpha in cell culture supernates (50 ul), serum (50 ul).Spezifikation
4.70 - 300pg/mL | |
1.5pg/mL | |
Cell Culture Supernatant, Serum |