missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL3/MIP-1 alpha Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure mouse MIP-1 alpha levels in cell culture supernates and serum.

Marke:  R&D Systems™ MMA00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412196

  • 933.07 € / 192 Tests


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains E. coli-expressed recombinant mouse MIP-1 alpha and antibodies raised against the recombinant protein. Results obtained for naturally occurring mouse MIP-1 alpha showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values of natural mouse MIP-1 alpha
Spezifikation

Spezifikation

4.70 - 300pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
20302
2 Plates
1.5pg/mL
Cell Culture Supernates (50μL), Serum (50μL)
Natural and recombinant mouse MIP-1 alpha
Sufficient materials to run ELISAs on at least two 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL), Serum (50uL)
Mouse
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos