Learn More
ACROBiosystems Mouse EGF Protein, Fc Tag
350.80 € - 2564.46 €
Spezifikation
Zugriffsnummer | NP_034243.2 |
---|---|
Molekulargewicht | 32.5 kDa |
Quelle | Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession # P01132-1). |
Immunogen | Asn 977 - Arg 1029 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16407489
|
ACROBiosystems
EGF-M5265-1MG |
1 mg |
2564.46 €
1 Milligramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16497479
|
ACROBiosystems
EGF-M5265-100UG |
100 μg |
350.80 €
100 Mikrogramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Human epidermal growth factor (EGF) is also known as HOMG4 and URG,and is a growth factor that plays an important role in the regulation of cell growth, proliferation, and differentiation by binding to its receptor EGFR. Epidermal growth factor can be found in human platelets, macrophages, urine, saliva, milk, and plasma. EGF is the founding member of the EGF-family of proteins. Members of this protein family have highly similar structural and functional characteristics. All family members contain one or more repeats of the conserved amino acid sequence. The biological effects of salivary EGF include healing of oral and gastroesophageal ulcers, inhibition of gastric acid secretion, stimulation of DNA synthesis as well as mucosal protection from intraluminal injurious factors such as gastric acid, bile acids, pepsin, and trypsin and to physical, chemical and bacterial agents. Because of the increased risk of cancer by EGF, inhibiting it decreases cancer risk.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 32.5 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_034243.2 | |
Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession # P01132-1). | |
-20°C | |
EGF | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |
32.5 kDa | |
Asn 977 - Arg 1029 | |
EGF,URG,HOMG4 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |