missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibronectin-ELISA-Kit für die Maus

Artikelnummer. 18087414
Change view
Click to view available options
Menge:
96 Tests
Packungsgröße:
96 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18087414 96 Tests 96 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18087414 Lieferant Invitrogen™ Lieferanten-Nr. EEL096

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

Fibronectin is a disulfide-bonded dimer with a molecular weight range of 230-250 kDa. In the extracellular matrix of several connective tissues and vessels, fibronectin is present as an insoluble protein that is extensively cross-linked by interchain disulfide bonds forming high molecular mass polymers. Fibronectin is most abundant during embryonic development and tissue remodeling. Fibronectin is also present at high concentrations as a soluble plasma protein. Fibronectin is organized as a linear series of repeating modules which form domains for interaction with fibronectin itself, other matrix components (e.g. collagen and heparin) and receptors on cells (e.g. integrins). Fibronectin is present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. Fibronectin has been implicated in carcinoma development in lung cancer. Further, Fibronectin expression is increased especially in non-small cell lung carcinoma. The adhesion of lung carcinoma cells to Fibronectin enhances tumorgenecity and confers resistance to apoptosis induced by standard chemotherapeutic agents. The gene encoding Fibronectin has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some Fibronectin variants has not been determined.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P11276
Assay-Bereich 1,56 bis 100 ng
Assay-Empfindlichkeit: 0.94 ng/mL
Konjugat HRP
Produkttyp ELISA
Probentyp Plasma, Serum
Zur Verwendung mit (Geräte) Kolorimetrisches Mikrotiterplatten-Lesegerät
Gen-ID (Entrez) 14268
Gensymbol FN1
Interassay-VK <10%
Intraassay-VK <10
Kitinhalt Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Marker oder Farbstoff HRP
Menge 96 Tests
Kennzeichnung RUO
Probenvolumen Plasma 100 μL, Serum 100 μL
Lagerungsbedingungen Informationen zur komponentenspezifischen Lagertemperatur finden Sie in der Produktdokumentation.
Ziel Maus
Prüfzeit 1 h 20 min
Gesamte Assay-Zeit 3 h 30 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.