missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse Kappa Light Chain Recombinant Secondary Antibody, Invitrogen™

Rabbit Recombinant Monoclonal Secondary Antibody

Marke:  Invitrogen SA510215

325.85 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21653" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15879883

  • 343.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody reacts to the kappa light chain of mouse immunoglobulins. No cross reactivity with the lamda light chain, human IgG, rat IgG, or goat IgG. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Mouse Kappa Light Chain Recombinant Monoclonal antibody specifically detects Mouse Kappa Light Chain in Mouse samples. It is validated for ELISA, Western Blot.
Spezifikation

Spezifikation

Mouse Kappa Light Chain
Recombinant Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4
Peptide corresponding to Mouse IgG.
100 μg
Secondary
-20° C, Avoid Freeze/Thaw Cycles
IgG
ELISA, Western Blot
RM103
Unconjugated
Rabbit
Protein A
RUO
Mouse
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only