missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Isotype Control (MOPC-21), PE, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen MA110415
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10415 can be used as a negative control to measure the non-specific binding of Mouse IgG1 in human and rat tissues.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG1 | |
MOPC-21 | |
PE | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
not available | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
Control, Flow Cytometry | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |