missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2a, clone: ZX4, Isotype Control, Invitrogen™

Mouse Isotype Control

Marke:  Invitrogen SA112079

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11565000

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

SA1-12079 has been successfully used in FACS procedures to assess non-specific binding of mouse monoclonal antibodies in human samples.
Spezifikation

Spezifikation

Mouse IgG2a
ZX4
Unconjugated
Mouse
Protein A/G
RUO
Not Applicable
Liquid
Control, Flow Cytometry
1mg/mL
IgG2, IgG2a
White blood cells
100 μg
Isotype Control (Primary)
Store at -20°C, Avoid Freeze/Thaw Cycles
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote