Learn More
Rabbit anti-Mouse IgG2a (Kappa light chain) Recombinant Secondary Antibody, Invitrogen™
Rabbit Recombinant Monoclonal Secondary Antibody
Marke: Invitrogen SA510219
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts to the Fab region of mouse IgG2ak. No cross reactivity with mouse IgG2al, IgG1, IgG3, IgM, IgA, IgE, human IgG, rat IgG, or goat IgG. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Mouse IgG2a (Kappa light chain) | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4 | |
Peptide corresponding to Mouse IgG. | |
100 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
ELISA, Western Blot | |
RM107 | |
Unconjugated | |
Rabbit | |
Protein A | |
RUO | |
Mouse | |
Liquid |
For Research Use Only