missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-12 (total) ELISA Kits

Measure levels of total interleukin-12 (p40 + p70) in mouse serum, plasma and culture supernatant samples using 96-well plates.

Marke:  Invitrogen™ EMIL12TOT

Weitere Details : Netto-Gewicht : 0.10000kg

Artikelnummer. 10485335

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Assay IL-12 p40 + p70 (total mouse interleukin-12) levels in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Mouse IL-12 (total) ELISA Kits.

These mouse IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of both active (p70) and inactive (p40) forms of interleukin-12 (IL-12 total) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Total (p70 and p40) natural and recombinant mouse interleukin-12 (IL-12)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 47 to 3000pg/mL

  • Sensitivity: < 12pg/mL

  • Precision: Dependent upon sample type (generally, intra- and inter-assay CV < 10%)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse G-CSF, GM-CSF, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MCP-1, MIP-1α, RANTES or TNFα, human IL-12, or pig IL-12

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

47 to 3000pg/mL
Mouse IL-12 Total (p70, p40) ELISA Kit
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
96 microplate wells
Absorbance at 450nm minus absorbance at 550nm
1 Plate Kit
3 hr.
<12pg/mL
Colorimetric ELISA (TMB substrate) with Plates
No significant cross-reaction with mouse G-CSF, GM-CSF, IFNg, IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MCP-1, MIP-1a, RANTES or TNFa, human IL-12, or pig IL-12
TMB Substrate Solution, 12mL, 30X Wash Buffer, 50mL, Adhesive Plate Covers, 6 ea, Anti-Mouse IL-12 Total Precoated 96-well Strip Plate, 1 ea, Biotinylated Antibody Reagent, 12mL, HRP Dilution Buffer, 13mL, Lyophilized Recombinant Mouse IL-12 Standard, 2 vials, Standard Diluent, 20mL, Streptavidin-HRP Concentrate, 75uL, Stop Solution, 13mL
Dependent upon sample type (generally, intra- and inter-assay CV <10%)
Total (p70 and p40) natural and recombinant mouse interleukin-12 (IL-12)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers