missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-13 Culture Media Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure mouse IL-13 levels in cell culture supernates.

Marke:  R&D Systems™ SM1300CB

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412806

  • 2733.67 € / 6 Stück


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains E. coli-expressed recombinant mouse IL-13 and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate the recombinant mouse IL-13 accurately. Results obtained using natural mouse IL-13 showed dose curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural mouse IL-13
Spezifikation

Spezifikation

7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
16163
1 Pk.
Cell Culture Supernates (50uL)
Mouse
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
6 Plates
1.5pg/mL
Cell Culture Supernates (50μL)
Natural and recombinant mouse IL-13
Sufficient materials to run ELISAs on at least six 96-well plates
Videos