missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-15 ELISA Development Kit (ABTS), PeproTech®

Artikelnummer. 30089175
Change view
Click to view available options
Menge:
10 × 96 Tests
Packungsgröße:
10 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30089175 10 × 96 Tests 10 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30089175 Lieferant Invitrogen™ Lieferanten-Nr. 900K188K

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P48346
Assay-Bereich Lot-specific
Assay-Empfindlichkeit: Lot-specific
Konjugat HRP
Produkttyp ELISA
Zur Verwendung mit (Geräte) Colorimetric Microplate Reader
Gen-ID (Entrez) 16168
Gensymbol IL15
Kitinhalt Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
Marker oder Farbstoff HRP
Menge 10 × 96 Tests
Kennzeichnung RUO
Lagerungsbedingungen Refer to product documentation for component specific storage temperature.
Ziel IL-15
Prüfzeit 1 hrs 45 mins
Gesamte Assay-Zeit 24 hrs 30 mins
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.