missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-5 DuoSet, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Mouse IL-5

Marke:  R&D Systems™ DY405

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413695

  • 666.85 € / 15 Stück


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation

Spezifikation

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
1 Kit
31.20 - 2000pg/mL
Biotin
16191
15 Plates
Mouse
Videos