missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat Leptin Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure soluble mouse leptin in cell culture supernates, serum*, and plasma.

Marke:  R&D Systems™ MOB00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412206

  • 933.07 € / 192 Tests


Voraussichtlicher Liefertermin 06-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains recombinant mouse leptin expressed in E. coli and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate the recombinant mouse leptin accurately. Results obtained measuring natural mouse leptin showed dose-response curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural mouse leptin. *Pregnant mouse serum from the second trimester of pregnancy through post-partum day 1 require acid/urea activation of mouse leptin
Spezifikation

Spezifikation

62.50 - 4000pg/mL
Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules
25608
22pg/mL
Cell Culture Supernates (50μL), Serum (20μL), EDTA Plasma (20μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
1 Kit
Cell Culture Supernates (50uL), Serum (20uL), EDTA Plasma (20uL)
Mouse, Rat
Sufficient materials to run ELISAs on at least two 96-well plates
Videos