Learn More
ACROBiosystems Mouse ROR1 Protein, Fc Tag
423.38 € - 2685.44 €
Spezifikation
Molekulargewicht | 68.4 kDa |
---|---|
Quelle | Mouse ROR1, Fc Tag (RO1-M5250) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139). |
Immunogen | Gln 30 - Glu 403 |
Lagerungsbedingungen | -20°C |
Gen-Alias | ROR1,NTRKR1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807607
|
ACROBiosystems
RO1-M5250-1MG |
1 mg |
2685.44 €
1 Milligramm |
Verfügbar ab: 02-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15897597
|
ACROBiosystems
RO1-M5250-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 02-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Tyrosine-protein kinase transmembrane receptor ROR1 is also known as Neurotrophic tyrosine kinase, receptor-related 1 (NTRKR1), which belongs to the protein kinase superfamily or tyr protein kinase family or ROR subfamily. ROR1 contains 1 FZ (frizzled) domain, 1 Ig-like C2-type (immunoglobulin-like) domain, 1 kringle domain, 1 protein kinase domain. ROR1 is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Isoform Short is strongly expressed in fetal and adult CNS and in a variety of human cancers, including those originating from CNS or PNS neuroectoderm. ROR1 could interact with casein kinase 1 epsilon (CK1ε) to activate phosphoinositide 3-kinase-mediated AKT phosphorylation and cAMP-response-element-binding protein (CREB), which was associated with enhanced tumor-cell growth.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 68.4 kDa. The protein migrates as 76-96 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
68.4 kDa | |
Gln 30 - Glu 403 | |
ROR1,NTRKR1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Mouse ROR1, Fc Tag (RO1-M5250) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139). | |
-20°C | |
ROR1 | |
Mouse | |
50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |