missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VEGF Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure mouse VEGF in cell culture supernates, tissue homogenates, mouse serum, and plasma.

Marke:  R&D Systems™ SMMV00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412826

  • 2733.67 € / 6 Stück


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

It contains Sf 21-expressed mouse VEGF and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate the recombinant mouse VEGF accurately. Results obtained using natural mouse VEGF showed dose-response curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural mouse VEGF
Spezifikation

Spezifikation

7.80 - 500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
22339
1 Pk.
Cell Culture Supernates (50uL), Tissue Homogenates (50uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL)
Mouse
HRP
ELISA
6 Plates
3pg/mL
Cell Culture Supernates (50μL), Tissue Homogenates (50μL), Serum (10μL), EDTA Plasma (10μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos