missing translation for 'onlineSavingsMsg'
Learn More

Moxidectin O-TBS, TRC

Artikelnummer. 30475275
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
50 mg
missing translation for 'unitSize'
50 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30475275 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30475275 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM74482050MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Moxidectin O-TBS
CAS 1646166-08-6
Summenformel C43 H67 N O8 Si
Menge 50 mg
Synonym (6R,23E,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-5-O-[(1,1-dimethylethyl)dimethylsilyl]-6,28-epoxy-23-(methoxyimino)-milbemycin B
InChI-Formel InChI=1S/C43H67NO8Si/c1-26(2)19-29(5)37-31(7)36(44-47-11)24-42(51-37)23-34-22-33(50-42)18-17-28(4)20-27(3)15-14-16-32-25-48-39-38(52-53(12,13)41(8,9)10)30(6)21-35(40(45)49-34)43(32,39)46/h14-17,19,21,26-27,31,33-35,37-39,46H,18,20,22-25H2,1-13H3/b15-14+,28-17+,29-19+,32-16-,44-36+/t27-,31-,33+,34-,35-,37+,38+,39+,42-,43+/m0/s1
SMILES CO\N=C\1/C[C@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C/4\CO[C@@H]5[C@H](O[Si](C)(C)C(C)(C)C)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)O[C@@H]([C@H]1C)\C(=C\C(C)C)\C
Molekulargewicht (g/mol) 754.08
Formelmasse 753.4636
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.