missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MPro N3

Description
MPro N3 is a coronavirus main protease (Mpro) inhibitor (respective IC50 values are 2.7, 4 and 8.8 μM for MHV-A29, HCoV-229E and FOPV replication in vitro). Inhibits viral cell infection in an MHV plaque reduction assay. Also inhibits SARS-CoV-2 viral cell entry in a plaque reduction assay (IC50 = 16.8 μM).
Specifications
Specifications
| Chemischer Name oder Material | N-[(5-Methyl-3-isoxazolyl)carbonyl]-L-alanyl-L-valyl-N-[(1 S,2 E)-4-oxo-1-[[(3 S)-2-oxo-3-pyrrolidinyl]methyl]-4-(phenylmethoxy)-2-buten-1-yl]-L-leucinamide |
| CAS | 884650-98-0 |
| Menge | 5 mg |
| Ziel | Additional Synthetic Protease Inhibitors |
| Summenformel | C35H48N6O8 |
| Reinheit | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?