Learn More
MRPL20 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561417
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ASQEHGLKYP ALIGNLVKCQ VELNRKVLAD LAIYEPKTFK SLAALASRRR HEGFAAALGD GKEPEGIFSR VVQYH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 21q. [provided by RefSeq, Jul 2008].Spezifikation
MRPL20 | |
Polyclonal | |
Unconjugated | |
MRPL20 | |
2610008D01Rik; 39S ribosomal protein L20, mitochondrial; 4930425I20Rik; L20mt; Mitochondrial large ribosomal subunit protein bL20m; mitochondrial ribosomal protein L20; MRPL20; MRP-L20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYC9 | |
MRPL20 | |
Recombinant protein corresponding to Human MRPL20 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only