Learn More
MRPL33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564294
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MFLSAVFFAK SKSKNILVRM VSEAGTGFCF NTKRNRLREK LTLLHYDPVV KQRVLFVE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
MRPL33 Polyclonal antibody specifically detects MRPL33 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MRPL33 | |
Polyclonal | |
Unconjugated | |
MRPL33 | |
39S ribosomal protein L33, mitochondrial; C2orf1; L33mt; Mitochondrial large ribosomal subunit protein bL33m; mitochondrial ribosomal protein L33; MRPL33; MRP-L33; RPL33L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75394 | |
MRPL33 | |
Recombinant protein corresponding to Human MRPL33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only