Learn More
MRPL37 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555274
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LTKTKLIEGL PEKVLSLVDD PRNHIENQDE CVLNVISHAR LWQTTEEIPK RETYCPVIVD NLIQLCKSQI LKHP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
MRPL37 Polyclonal antibody specifically detects MRPL37 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
MRPL37 | |
Polyclonal | |
Unconjugated | |
MRPL37 | |
2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZE1 | |
MRPL37 | |
Recombinant protein corresponding to Human MRPL37 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only