Learn More
MRPS7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554663
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LKGGRFYQVP VPLPDRRRRF LAMKWMITEC RDKKHQRTLM PEKLSHKLLE AFHNQGPVIK RKHDLHKMAE ANRALAHYRW W Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
MRPS7 Polyclonal antibody specifically detects MRPS7 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
MRPS7 | |
Polyclonal | |
Unconjugated | |
MRPS7 | |
28S ribosomal protein S7, mitochondrial; 30S ribosomal protein S7 homolog; bMRP27a; bMRP-27a; mitchondrial ribosomal protein S7; mitochondrial ribosomal protein S7; Mitochondrial small ribosomal subunit protein uS7m; MRP-S; Mrps7; MRP-S7; ribosomal protein, mitochondrial, S7; Rpms7; RP-S7; S7mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51081 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2R9 | |
MRPS7 | |
Recombinant protein corresponding to Human MRPS7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only