Learn More
MSMP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563079
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FTLGESWLRK DCFHCTCLHP VGVGCCDTSQ HPIDFPAGCE VRQEAGTCQF SLVQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
MSMP Polyclonal antibody specifically detects MSMP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MSMP | |
Polyclonal | |
Unconjugated | |
MSMP | |
microseminoprotein, prostate associated; MSMP; PC3 prostate cancer cell-secreted microprotein; PC3-secreted microprotein; Prostate-associated microseminoprotein; PSMP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
692094 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q1L6U9 | |
MSMP | |
Recombinant protein corresponding to Human MSMP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only