Learn More
MSTO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552227
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DRACHTSQLT PGTPPPSALH ACTTGEEILA QYLQQQQPGV MSSSHLLLTP CRVAPPYPHL FSSCSPPGMV LDGSPKGAAV ESIPVFGALC SSSSLHQTLE ALARDLTKLD LRRWASFMDA GVEHDDVAEL LQELQSLAQC YQGGDSLVD Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
MSTO1 Polyclonal antibody specifically detects MSTO1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MSTO1 | |
Polyclonal | |
Unconjugated | |
MSTO1 | |
LST005; misato 1, mitochondrial distribution and morphology regulator; misato homolog 1; MST; MSTO1; Protein misato homolog 1; SLTP005 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUK6 | |
MSTO1 | |
Recombinant protein corresponding to Human MSTO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only