Learn More
MT-ND5 Polyclonal Antibody, Invitrogen™
Artikelnummer. 15506191
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15506191 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15506191 Lieferant Invitrogen Lieferanten-Nr. PA539277
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 5 (ND5) is most likely a component of the hydrophobic protein fragment of Complex I. Mutations in the gene encodiing for ND5 are implicated in Mitochondrial neurogastrointestinal encephalomyopathy, an autosomal recessive multisystem disorder that results in dramatically elevated levels of circulating thymidine and deoxyuridine.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen MT-ND5
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gen MT-ND5
Gen-Zugriffsnummer P03915
Gen-Alias NADH-ubiquinone oxidoreductase chain 5, NADH dehydrogenase subunit 5, MT-ND5, MTND5, NADH5, ND5
Gensymbole MT-ND5
Wirtsspezies Rabbit
Immunogen A synthetic peptide derived from the internal region of human MT-ND5
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 4540
Zielspezies Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG