missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16957-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Transcription termination factor. Binds to a 28 bp region within the tRNA(Leu(uur)) gene at a position immediately adjacent to and downstream of the 16S rRNA gene; this region comprises a tridecamer sequence critical for directing accurate termination. Binds DNA along the major grove and promotes DNA bending and partial unwinding. Promotes base flipping. Transcription termination activity appears to be polarized with highest specificity for transcripts initiated on the light strand.Spezifikation
MTERF | |
Polyclonal | |
Unconjugated | |
MTERF1 | |
MTERF, MTERF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
545725, 7978, 85261 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CHZ9, Q99551, Q9EPI8 | |
Mterf1, Mterf1a | |
MTERF Fusion Protein Ag10640 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |