Learn More
MTFMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559261
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RENKEEELID KLEVVTMPSP SPKGLPVKQY AVQSQLPVYE WPDVGSGEYD VGVVASFGRL LNEALILKFP YGILNVHPSC LPRWRGPAPV IH Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.Spezifikation
MTFMT | |
Polyclonal | |
Unconjugated | |
MTFMT | |
2310020P08Rik; COXPD15; Fmt; FMT1; LOW QUALITY PROTEIN: methionyl-tRNA formyltransferase, mitochondrial; methionyl-tRNA formyltransferase, mitochondrial; mitochondrial methionyl-tRNA formyltransferase; mitochondrial methionyl-tRNA transformylase; MtFMT; RGD1309462 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123263 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DP5 | |
MTFMT | |
Recombinant protein corresponding to Human MTFMT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only