Learn More
MTHFS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552509
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VLPKTSWNIP QPGEGDVREE ALSTGGLDLI FMPGLGFDKH GNRLGRGKGY YDAYLKRCLQ HQEVKPYTLA LAFKEQICLQ VPVNENDMKV DEVLYEDSST A Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
MTHFS Polyclonal antibody specifically detects MTHFS in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
MTHFS | |
Polyclonal | |
Unconjugated | |
Mthfs | |
1110034I12Rik; 2310020H23Rik; 5, 10-methenyltetrahydrofolate synthetase; 5,10-methenyl-tetrahydrofolate synthetase; 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase); 5-formyltetrahydrofolate cyclo-ligase; AI119695; FLJ30410; HsT19268; methenyl-THF synthetase; Mthfs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49914 | |
Mthfs | |
Recombinant protein corresponding to Human MTHFS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only