Learn More
MTMR12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562384
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RPEEIHTNEK EVTEKEVTLH LLPGEQLLCE ASTVLKYVQE DSCQHGVYGR LVCTDFKIAF LGDDESALDN DETQFKNKVI GENDITLHCV DQIYGVFDE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
MTMR12 Polyclonal antibody specifically detects MTMR12 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MTMR12 | |
Polyclonal | |
Unconjugated | |
MTMR12 | |
3Pap; 3-PAP; 3-phosphatase adapter protein; 3-phosphatase adapter subunit; 4932703C11; C730015A02Rik; im:7160376; Inactive phosphatidylinositol 3-phosphatase 12; Kiaa1682; mKIAA1682; Mtmr12; myotubularin related protein 12; myotubularin-related protein 12; Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol 3-phosphatase-associated protein; phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit; phosphatidylinositol-3-phosphatase associated protein; phosphatidylinositol-3-phosphatase-associated protein; phosphatidylinositol-3-phosphate associated protein; Pip3ap; si:dkey-207e22.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54545 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0I1 | |
MTMR12 | |
Recombinant protein corresponding to Human MTMR12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only