Learn More
MTO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556409
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MSCNPSFGGI GKGHLMREVD ALDGLCSRIC DQSGVHYKVL NRRKGPAVWG LRAQIDRKLY KQNMQKEILN TPLLTVQEGA VEDLILTEPE PEHTGKCRVS GVVL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
MTO1 Polyclonal antibody specifically detects MTO1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MTO1 | |
Polyclonal | |
Unconjugated | |
MTO1 | |
2310039H01Rik; 5730419A02Rik; CGI02; CGI-02; COXPD10; homolog of yeast Mto1; LOW QUALITY PROTEIN: protein MTO1 homolog, mitochondrial; mitochondrial MTO1-3; mitochondrial translation optimization 1 homolog; mitochondrial translation optimization 1 homolog (S. cerevisiae); mitochondrial tRNA translation optimization 1; MTO1; Protein MTO1 homolog, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25821 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2Z2 | |
MTO1 | |
Recombinant protein corresponding to Human MTO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only