Learn More
MTX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552838
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: WIDTKNYVEV TRKWYAEAMP FPLNFFLPGR MQRQYMERLQ LLTGEHRPED EEELEKELYR EARECLTLLS QRLGSQKFFF GDAPASLDAF VFSYLALLLQ AKLPSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
MTX1 Polyclonal antibody specifically detects MTX1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MTX1 | |
Polyclonal | |
Unconjugated | |
MTX1 | |
Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13505 | |
MTX1 | |
Recombinant protein corresponding to Human MTX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only