Learn More
MUC7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552292
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RRHHHQSPKS HFELPHYPGL LAHQKPFIRK SYKCLHKRCR PKLPPSPNNP PKFPNPHQPP KHPDKNSSVV NPTLVATTQI PSVTFPSAST KITTLPNVTF LPQNATTISS RENVNTSSSV ATLAPVNSPA P Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.
MUC7 Polyclonal antibody specifically detects MUC7 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MUC7 | |
Polyclonal | |
Unconjugated | |
MUC7 | |
apo-MG2; MG2; MUC7; MUC-7; mucin 7, salivary; mucin 7, secreted; mucin-7; Salivary mucin-7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAX7 | |
MUC7 | |
Recombinant protein corresponding to Human MUC7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only