Learn More
TRPML2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553926
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YHQLKDITLG TLGYGENEDN RIGLKVCKQH YKKGTMFPSN ETLNIDNDVE LDCVQLDLQD LSKKPPDWKN SSFFRLEFYR Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 66%.
TRPML2 Polyclonal antibody specifically detects TRPML2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TRPML2 | |
Polyclonal | |
Unconjugated | |
MCOLN2 | |
3300002C04Rik; AI549968; C86638; FLJ36691; MCLN2; MCOLN2; mucolipidin 2; mucolipin 2; mucolipin-2; Transient receptor potential channel mucolipin 2; TRPML2; TRP-ML2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZK6 | |
MCOLN2 | |
Recombinant protein corresponding to Human Mucolipin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only