missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Spezifikation
Spezifikation
| Antigen | Muscle Phosphofructokinase/PFKM/PFK-1 |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konzentration | 1.0 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate |
| Zusammensetzung | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gen-Zugriffsnummer | NP_000280.1 |
| Gen-Alias | 6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase |
| Gensymbole | PFKM |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?