missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUSK Recombinant Human Protein
MUSK Recombinant Human Protein
Marke: Invitrogen PV3834
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn.Spezifikation
NP_005583.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
69.4kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MUSK | |
MUSK was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 5mM MnCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 500μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 519-869 | |
29nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
80% by SDS-PAGE |
0.16mg/mL | |
4593 | |
MUSK Recombinant Human Protein | |
–80°C | |
1 Each | |
MUSK Recombinant Human Protein | |
MUSK | |
MUSK Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.