Learn More
MYH1 Mouse anti-Human, Unconjugated, Clone: 1G10H9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67299-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The MYH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYH1. This antibody recognizes human, mouse, pig antigen. The MYH1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
| MYH1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G10H9 | |
| Unconjugated | |
| 150 μL | |
| 4619 | |
| IgG2a |