missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications
Specifications
| Antigen | MYL3 |
| Anwendungen | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.3 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gen | MYL3 |
| Gen-Zugriffsnummer | P08590, P09542, P16409 |
| Gen-Alias | Cardiac myosin light chain 1, CMH8, CMLC1, MLC1SB, MLC1V, MYL3, Myosin light chain 3, VLC1 |
| Gensymbole | Myl3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?