missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLIP Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15455-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.Spezifikation
MYLIP | |
Polyclonal | |
Unconjugated | |
MYLIP | |
BZF1, Idol, MIR, MYLIP, PP5242 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100760940, 218203, 29116, 306825 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZDI6, Q8BM54, Q8WY64 | |
MYLIP | |
MYLIP Fusion Protein Ag7720 | |
150 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |