missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Brom-4-Fluorophenyl)-2-Methoxybenzamid, 97 %, Thermo Scientific™

Catalog Number p-7039882
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1021439-52-0
C14H11BrFNO2
324.149
MFCD12559362
CVEPLYNHBLSXCV-UHFFFAOYSA-N
n-2-bromo-4-fluorophenyl-2-methoxybenzamide
28938569
N-(2-Brom-4-Fluorphenyl)-2-methoxybenzamid
COC1=CC=CC=C1C(=O)NC2=C(C=C(C=C2)F)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1021439-52-0
C14H11BrFNO2
324.149
MFCD12559362
CVEPLYNHBLSXCV-UHFFFAOYSA-N
n-2-bromo-4-fluorophenyl-2-methoxybenzamide
28938569
N-(2-Brom-4-Fluorphenyl)-2-methoxybenzamid
COC1=CC=CC=C1C(=O)NC2=C(C=C(C=C2)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(2-Bromo-4-fluorophenyl)-2-methoxybenzamide
Menge 250 mg
Formelmasse 324.15
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'