missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Ethoxyphenyl)-2-Methoxybenzamid, 97 %, Thermo Scientific™

Catalog Number p-7040137
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
306279-65-2
C16H17NO3
271.316
MFCD01010708
IFOHCGGZEGVBME-UHFFFAOYSA-N
n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
700574
N-(2-ethoxyphenyl)-2-methoxybenzamid
CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
306279-65-2
C16H17NO3
271.316
MFCD01010708
IFOHCGGZEGVBME-UHFFFAOYSA-N
n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
700574
N-(2-ethoxyphenyl)-2-methoxybenzamid
CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(2-Ethoxyphenyl)-2-methoxybenzamide
Menge 250 mg
Formelmasse 271.32
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'