missing translation for 'onlineSavingsMsg'
Learn More

N-(2-hydroxyphenyl)thioharnstoff, 97 %, Thermo Scientific Chemicals

Catalog Number p-7045386
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1520-26-9
C7H8N2OS
168.214
MFCD00041159
VYBCFZXLXJUFPM-UHFFFAOYSA-N
1-2-hydroxyphenyl thiourea, 2-hydroxyphenyl thiourea, 2-hydroxy-phenyl-thiourea, 2-aminothioxomethyl amino phenol, o-hydroxyphenylthiourea, acmc-20ant0, thiourea, hydroxyphenyl, 1-o-hydroxyphenyl thiourea, n-2-hydroxyphenyl thiourea, thiourea, 2-hydroxyphenyl
3003569
(2-hydroxyphenyl)thioharnstoff
C1=CC=C(C(=C1)NC(=S)N)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1520-26-9
C7H8N2OS
168.214
MFCD00041159
VYBCFZXLXJUFPM-UHFFFAOYSA-N
1-2-hydroxyphenyl thiourea, 2-hydroxyphenyl thiourea, 2-hydroxy-phenyl-thiourea, 2-aminothioxomethyl amino phenol, o-hydroxyphenylthiourea, acmc-20ant0, thiourea, hydroxyphenyl, 1-o-hydroxyphenyl thiourea, n-2-hydroxyphenyl thiourea, thiourea, 2-hydroxyphenyl
3003569
(2-hydroxyphenyl)thioharnstoff
C1=CC=C(C(=C1)NC(=S)N)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(2-Hydroxyphenyl)thiourea
Schmelzpunkt ∽158°C (decomposition)
Geruch Odorless
Menge 1 g
Beilstein 1102479
Formelmasse 168.22
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'