Learn More
N-(3-((4-Amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino)propyl)tetrahydrofuran-2-carboxamide hydrochloride, 98%, BLD Pharm™
Product Code. 31448124
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31448124 5 g
31445913 1 g
31439325 250 mg
31438079 100 mg
31436710 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31448124 Supplier BLD Pharm Supplier No. BD1129345g
 Restricted Item
Verfügbar ab: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 81403-68-1
Molecular Formula C19H28ClN5O4
MDL Number MFCD00879135
Quantity 5 g
SMILES O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C.Cl
Molecular Weight (g/mol) 425.91
Percent Purity 98%
Chemical Name or Material N-(3-((4-Amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino)propyl)tetrahydrofuran-2-carboxamide hydrochloride