missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Desethinylphenyl)-N-(3-bromphenyl) Erlotinibhydrochlorid, TRC
Specifications
Specifications
| Chemischer Name oder Material | N-(3-Desethynylphenyl)-N-(3-bromophenyl) Erlotinib hydrochloride |
| CAS | 2055840-41-8 |
| Summenformel | C20 H22 Br N3 O4 . Cl H |
| Menge | 100 mg |
| Synonym | 4-Quinazolinamine, N-(3-bromophenyl)-6,7-bis(2-methoxyethoxy)-, hydrochloride (1:1), N-(3-Bromophenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine hydrochloride, N-(3-Bromophenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride, N-(3-Desethynylphenyl)-N-(3-bromophenyl) Erlotinib hydrochloride, N-(3-Desethynylphenyl)-N-(3-bromophenyl)erlotinib hydrochloride, Erlotinib impurity 3 hydrochloride |
| InChI-Formel | InChI=1S/C20H22BrN3O4.ClH/c1-25-6-8-27-18-11-16-17(12-19(18)28-9-7-26-2)22-13-23-20(16)24-15-5-3-4-14(21)10-15;/h3-5,10-13H,6-9H2,1-2H3,(H,22,23,24);1H |
| SMILES | Cl.COCCOc1cc2ncnc(Nc3cccc(Br)c3)c2cc1OCCOC |
| IUPAC-Name | N-(3-Bromphenyl)-6,7-bis(2-methoxyethoxy)chinazolin-4-amin;hydrochlorid |
| Molekulargewicht (g/mol) | 484.77 |
| Formelmasse | 483.056 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?