missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromphenyl)Benzenesulfonamid, 97 %, Thermo Scientific Chemicals

Catalog Number p-7036975
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16468-97-6
C12H10BrNO2S
312.181
MFCD00159375
HHLFFQAEABRCOJ-UHFFFAOYSA-N
n-4-bromophenyl benzenesulfonamide, benzenesulfonamide, n-4-bromophenyl, 4'-bromobenzenesulfonanilide, benzenesulfonanilide, 4'-bromo, 4-bromophenyl phenylsulfonyl amine, 4/'-bromobenzenesulfonanilide, n-p-bromophenylbenzenesulphonamide, n-4-bromophenyl benzenesulfonamide #
293101
N-(4-Bromphenyl)benzolsulfonamid
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16468-97-6
C12H10BrNO2S
312.181
MFCD00159375
HHLFFQAEABRCOJ-UHFFFAOYSA-N
n-4-bromophenyl benzenesulfonamide, benzenesulfonamide, n-4-bromophenyl, 4'-bromobenzenesulfonanilide, benzenesulfonanilide, 4'-bromo, 4-bromophenyl phenylsulfonyl amine, 4/'-bromobenzenesulfonanilide, n-p-bromophenylbenzenesulphonamide, n-4-bromophenyl benzenesulfonamide #
293101
N-(4-Bromphenyl)benzolsulfonamid
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(4-Bromophenyl)benzenesulfonamide
Menge 250 mg
Formelmasse 312.18
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'