missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-9-(acetylamino)-9-desoxyneuraminsäure, TRC

Artikelnummer. 30316925
Change view
Click to view available options
Menge:
1 mg
Unit Size:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30316925 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316925 Supplier Toronto Research Chemicals Supplier No. TRCA1680051MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material N-Acetyl-9-(acetylamino)-9-deoxyneuraminic Acid
CAS 112054-78-1
Summenformel C13H22N2O9
Menge 1 mg
Synonym 5,9-Bis(acetylamino)-3,5,9-trideoxy-D-glycero-D-galacto-2-nonulosonic Acid, 9-Acetamido-9-N-(acetyl)-9-deoxy-D-neuraminic Acid
InChI-Formel nChI=1S/C13H22N2O9/c1-5(16)14-4-8(19)10(20)11-9(15-6(2)17)7(18)3-13(23,24-11)12(21)22/h7-11,18-20,23H,3-4H2,1-2H3,(H,14,16)(H,15,17)(H,21,22)/t7?,8-,9?,10-,11?,13?/m1/s1
SMILES O[C@@H]1[C@@H](NC(C)=O)[C@H]([C@H](O)[C@H](O)CNC(C)=O)OC(O)(C(O)=O)C1
IUPAC-Name 5-Acetamido-6-[(1R,2R)-3-Acetamido-1,2-diHydroxypropyl]-2,4-DiHydroxyoxan-2-Carbonsäure
Molekulargewicht (g/mol) 350.32
Formelmasse 350.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.