Learn More
N-Acetyl-DL-Norvalin, 98 %, Thermo Scientific™
Catalog Number p-7028207
Click to view available options
Menge:
1 g
5 g
25 g
7682-15-7
C7H13NO3
159.19
MFCD00037269
BSYFPUSAWVWWDG-UHFFFAOYSA-N
n-acetyl-dl-norvaline, ac-dl-nva-oh, acetyl-dl-norvaline, n-acetylnorvaline #, 2-acetamido-pentanoic acid, 2-acetylamino valeric acid, n-alpha-acetyl-dl-norvaline, n-acetyl-2-aminovaleric acid, 2-acetylamino pentanoic acid
306106
2-Acetamidopentansäure
CCCC(NC(C)=O)C(O)=O
7682-15-7
C7H13NO3
159.19
MFCD00037269
BSYFPUSAWVWWDG-UHFFFAOYSA-N
n-acetyl-dl-norvaline, ac-dl-nva-oh, acetyl-dl-norvaline, n-acetylnorvaline #, 2-acetamido-pentanoic acid, 2-acetylamino valeric acid, n-alpha-acetyl-dl-norvaline, n-acetyl-2-aminovaleric acid, 2-acetylamino pentanoic acid
306106
2-Acetamidopentansäure
CCCC(NC(C)=O)C(O)=O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Acetyl-DL-norvaline
Formelmasse 159.19
Reinheit (%) 98%
Menge 1 g