missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-Serin, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7030170
Click to view available options
Menge:
5 g
25 g
97-14-3
C5H9NO4
147.13
MFCD00037248
JJIHLJJYMXLCOY-UHFFFAOYSA-N
n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l
352294
2-amino-3-hydroxypropansäure
CC(=O)NC(CO)C(=O)O
97-14-3
C5H9NO4
147.13
MFCD00037248
JJIHLJJYMXLCOY-UHFFFAOYSA-N
n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l
352294
2-amino-3-hydroxypropansäure
CC(=O)NC(CO)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Acetyl-DL-serine
Formelmasse 147.13
Reinheit (%) ≥98%
Menge 5 g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'