Learn More
N-Acetylglufosinat-d3-Dinatriumsalz, TRC
Specifications
Specifications
| Chemischer Name oder Material | N-Acetyl Glufosinate-d3 Sodium |
| CAS | 1356933-74-8 |
| Summenformel | C7H9D3NNa2O5P |
| Menge | 50 mg |
| Synonym | 2-(Acetyl-d3-amino)-4-(hydroxymethylphosphinyl)butanoic acid disodium salt, 4-[Hydroxy(methyl)phosphoryl]-2-trideuterioacetamidobutanoic acid disodium salt, N-(Acetyl-d3) glufosinate disodium, N-(Acetyl-d3)-DL-glufosinate disodium, Glufosinate-N-acetyl-d3 disodium salt, Glufosinate-N-acetyl-D3 (acetyl-D3) disodium, N-Acetyl glufosinate-d3 disodium salt |
| InChI-Formel | InChI=1S/C7H14NO5P.2Na/c1-5(9)8-6(7(10)11)3-4-14(2,12)13;;/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)(H,12,13);;/q;2*+1/p-2/i1D3;; |
| SMILES | [Na+].[Na+].[2H]C([2H])([2H])C(=O)NC(CCP(=O)(C)[O-])C(=O)[O-] |
| IUPAC-Name | Dinatrium;4-[methyl(oxido)phosphoryl]-2-[(2,2,2-trideuterioacetyl)amino]butanoat |
| Molekulargewicht (g/mol) | 270.15 |
| Formelmasse | 270.0437 |