missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals n-Acetyl-Leu-Glu-His-Asp-7-Amino-4-Methylcumarin
Click to view available options
Menge:
1mg
5mg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Farbe | Weiß |
Inhalt und Lagerung | Store at -20°C |
Format | Lyophilisiertes Pulver |
Molekulargewicht | 711.73 |
Produkttyp | N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin |
Menge | 1mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'