missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminsäure, 99 %, Thermo Scientific Chemicals

Catalog Number p-50156
Click to view available options
Menge:
25 mg
100 mg
Verpackung:
Glasflasche
131-48-6
C11H19NO9
309.271
MFCD00006620
SQVRNKJHWKZAKO-LFIUDZTESA-N
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
126963458
(4S,5S,6R)-5-Acetamido-2,4-Dihydroxy-6-[(1R,2r)-1,2,3-Trihydroxypropyl]oxan-2-Carbonsäure
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
131-48-6
C11H19NO9
309.271
MFCD00006620
SQVRNKJHWKZAKO-LFIUDZTESA-N
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
126963458
(4S,5S,6R)-5-Acetamido-2,4-Dihydroxy-6-[(1R,2r)-1,2,3-Trihydroxypropyl]oxan-2-Carbonsäure
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Acetylneuraminic acid
Namenshinweis 98%
CAS Min. % 98.5
CAS Max. % 100.0
Farbe Weiß
Infrarotspektrum Echt
Prozentgehaltsbereich 99%
Verpackung Glasflasche
Menge 25 mg
Merck Index 15, 8623
Spezifische Drehung -30° to -34° (20°C, 589nm) (c=2, water)
Formelmasse 309.26
Reinheit (%) ≥98.5%
Physikalische Form Pulver
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'